Product Details

SNP ID
rs9914481
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7250530 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACAGGGACACCCAAACTTCAATG[C/G]TGGGCACAGCACCCACCTTGAACTC
Phenotype
MIM: 609131 MIM: 610684 MIM: 615019 MIM: 605125
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CLDN7 PubMed Links

Gene Details

Gene
CLDN7
Gene Name
claudin 7
There are no transcripts associated with this gene.

Gene
CTDNEP1
Gene Name
CTD nuclear envelope phosphatase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143775.1 Intron NP_001137247.1
NM_015343.4 Intron NP_056158.2
Gene
ELP5
Gene Name
elongator acetyltransferase complex subunit 5
There are no transcripts associated with this gene.

Gene
GABARAP
Gene Name
GABA type A receptor-associated protein
There are no transcripts associated with this gene.

View Full Product Details