Product Details

SNP ID
rs9690073
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:40154244 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGAACTTGCTTTGAGAGCAAAATA[A/G]TATTTAAAAAAGTGCAGTGTGCACT
Phenotype
MIM: 609187
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SUGCT PubMed Links

Gene Details

Gene
SUGCT
Gene Name
succinyl-CoA:glutarate-CoA transferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193311.1 Intron NP_001180240.1
NM_001193312.1 Intron NP_001180241.1
NM_001193313.1 Intron NP_001180242.1
NM_024728.2 Intron NP_079004.1
XM_006715775.3 Intron XP_006715838.1
XM_011515525.2 Intron XP_011513827.1
XM_011515526.2 Intron XP_011513828.1
XM_011515527.2 Intron XP_011513829.1
XM_011515528.2 Intron XP_011513830.1
XM_011515529.2 Intron XP_011513831.1
XM_011515530.2 Intron XP_011513832.1
XM_017012621.1 Intron XP_016868110.1
XM_017012622.1 Intron XP_016868111.1

View Full Product Details