Product Details

SNP ID
rs9323925
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:95416583 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAGTCACCGCGGCCCCAGCCCAGG[C/T]GGTGCATGGAGCCCAGCTCGCCTAC
Phenotype
MIM: 610861
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00341 PubMed Links

Gene Details

Gene
LINC00341
Gene Name
long intergenic non-protein coding RNA 341
There are no transcripts associated with this gene.

Gene
SYNE3
Gene Name
spectrin repeat containing nuclear envelope family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152592.3 4170 Intron NP_689805.3
XM_005267377.3 4170 UTR 3 XP_005267434.1
XM_006720063.3 4170 UTR 3 XP_006720126.1
XM_011536515.2 4170 UTR 3 XP_011534817.1
XM_017021056.1 4170 UTR 3 XP_016876545.1
XM_017021057.1 4170 Intron XP_016876546.1

View Full Product Details