Product Details
- SNP ID
-
rs8004831
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:36522806 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGCCGAAAATAAGCCCGGAGACTAA[C/G]GCACCTTGTAGGGAAGAGAAGGACA
- Phenotype
-
MIM: 600635
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
NKX2-1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs192994917] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NKX2-1
- Gene Name
- NK2 homeobox 1
There are no transcripts associated with this gene.
- Gene
- NKX2-1-AS1
- Gene Name
- NKX2-1 antisense RNA 1
There are no transcripts associated with this gene.
- Gene
- SFTA3
- Gene Name
- surfactant associated 3
There are no transcripts associated with this gene.
View Full Product Details