Product Details

SNP ID
rs6703669
Assay Type
Functionally tested
NCBI dbSNP Submissions
34
Location
Chr.1:46396883 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGCTGGCTGTAATGTATGAATTTA[C/T]TCCAAACCATCCCTGGGGCCAAGAT
Phenotype
MIM: 602935
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAAH PubMed Links

Gene Details

Gene
FAAH
Gene Name
fatty acid amide hydrolase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001441.2 Intron NP_001432.2

View Full Product Details