Product Details

SNP ID
rs10057858
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:157625217 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGTTGGTGACTATTTCCCATCAT[C/T]AATTCACTTTGTACCTTAGAGATCT
Phenotype
MIM: 606698
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SOX30 PubMed Links

Gene Details

Gene
SOX30
Gene Name
SRY-box 30
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308165.1 2491 Intron NP_001295094.1
NM_007017.2 2491 Intron NP_008948.1
NM_178424.1 2491 Intron NP_848511.1
XM_005265803.4 2491 UTR 3 XP_005265860.1
XM_011534420.1 2491 UTR 3 XP_011532722.1

View Full Product Details