Product Details
- SNP ID
-
rs8043156
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:50526576 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAAAAAGGTATTCTGTTTGCTAATG[C/T]ATATGTACAAGCATGTGTTAACTGT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
USP50
PubMed Links
Gene Details
- Gene
- USP50
- Gene Name
- ubiquitin specific peptidase 50
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_203494.4 |
|
Intron |
|
|
NP_987090.2 |
View Full Product Details