Product Details

SNP ID
rs9810328
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:184710370 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCATCACAGAAAACAAGATGCTCC[A/G]CAAATGTTTAATTTATTAAGTGTGC
Phenotype
MIM: 609267
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAGEF1 PubMed Links

Gene Details

Gene
MAGEF1
Gene Name
MAGE family member F1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022149.4 1679 UTR 3 NP_071432.2

View Full Product Details