Product Details

SNP ID
rs9285274
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:71439902 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCAACATTATTATCTTTAATTCTA[C/T]ATACCACTACCAGTTTGGAATGAAA
Phenotype
MIM: 603803
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DACH1 PubMed Links

Gene Details

Gene
DACH1
Gene Name
dachshund family transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004392.6 2323 UTR 3 NP_004383.4
NM_080759.5 2323 UTR 3 NP_542937.3
NM_080760.5 2323 UTR 3 NP_542938.3
XM_011534939.2 2323 UTR 3 XP_011533241.1
XM_011534940.2 2323 Intron XP_011533242.1
XM_011534941.2 2323 Intron XP_011533243.1
XM_011534942.2 2323 UTR 3 XP_011533244.1
XM_017020396.1 2323 UTR 3 XP_016875885.1

View Full Product Details