Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_015046.5 | 4196 | UTR 3 | NP_055861.3 | ||
XM_005272171.1 | 4196 | UTR 3 | XP_005272228.1 | ||
XM_005272172.2 | 4196 | Intron | XP_005272229.1 | ||
XM_005272173.2 | 4196 | Intron | XP_005272230.1 | ||
XM_011518404.2 | 4196 | Intron | XP_011516706.1 | ||
XM_011518405.2 | 4196 | Intron | XP_011516707.1 | ||
XM_011518406.2 | 4196 | Intron | XP_011516708.1 | ||
XM_011518407.1 | 4196 | Intron | XP_011516709.1 | ||
XM_011518408.2 | 4196 | Intron | XP_011516710.1 | ||
XM_017014495.1 | 4196 | UTR 3 | XP_016869984.1 | ||
XM_017014496.1 | 4196 | UTR 3 | XP_016869985.1 | ||
XM_017014497.1 | 4196 | UTR 3 | XP_016869986.1 |