Product Details

SNP ID
rs9261083
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30001428 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTATAACAACACCACATTCCACAT[C/G]TAGACATTTTGCTTTGGCCCACAGT
Phenotype
MIM: 615714
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HLA-J PubMed Links

Gene Details

Gene
HLA-J
Gene Name
major histocompatibility complex, class I, J (pseudogene)
There are no transcripts associated with this gene.

Gene
ZNRD1ASP
Gene Name
zinc ribbon domain containing 1 antisense, pseudogene
There are no transcripts associated with this gene.

View Full Product Details