Product Details

SNP ID
rs7413066
Assay Type
Functionally tested
NCBI dbSNP Submissions
48
Location
Chr.1:84481062 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTTCTATCTTATATTAGGGAATCC[C/T]TCCTGATATTTAAGTTGAATTACTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RPF1 PubMed Links

Gene Details

Gene
RPF1
Gene Name
ribosome production factor 1 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025065.6 Intron NP_079341.2

View Full Product Details