Product Details

SNP ID
rs7087360
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:16830338 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACCCACGATTATCTTTATAGAACG[A/C]TTTTCTTAAGATATTTAAAGTGAGT
Phenotype
MIM: 602997
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
CUBN PubMed Links

Gene Details

Gene
CUBN
Gene Name
cubilin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081.3 Intron NP_001072.2
XM_011519708.2 Intron XP_011518010.1
XM_011519709.2 Intron XP_011518011.1
XM_011519710.2 Intron XP_011518012.1
XM_011519711.2 Intron XP_011518013.1

View Full Product Details