Product Details

SNP ID
rs4768959
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:51051878 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCATAGCAGATTACAGAAGCCAAA[C/T]GTAGCTAGAAGAGAGGCGATGTGCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CSRNP2 PubMed Links

Gene Details

Gene
CSRNP2
Gene Name
cysteine and serine rich nuclear protein 2
There are no transcripts associated with this gene.

Gene
LETMD1
Gene Name
LETM1 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243689.1 Intron NP_001230618.1
NM_001300765.1 Intron NP_001287694.1
NM_015416.4 Intron NP_056231.3
XM_006719335.1 Intron XP_006719398.1
XM_006719336.1 Intron XP_006719399.1
XM_006719339.1 Intron XP_006719402.1
XM_006719340.1 Intron XP_006719403.1
XM_011538161.2 Intron XP_011536463.1
XM_011538162.2 Intron XP_011536464.1
XM_011538163.2 Intron XP_011536465.1
XM_011538164.2 Intron XP_011536466.1
XM_011538165.2 Intron XP_011536467.1
XM_011538167.1 Intron XP_011536469.1
XM_017019151.1 Intron XP_016874640.1
XM_017019152.1 Intron XP_016874641.1
XM_017019153.1 Intron XP_016874642.1
XM_017019154.1 Intron XP_016874643.1
XM_017019155.1 Intron XP_016874644.1
XM_017019156.1 Intron XP_016874645.1
XM_017019157.1 Intron XP_016874646.1
XM_017019158.1 Intron XP_016874647.1
XM_017019159.1 Intron XP_016874648.1
XM_017019160.1 Intron XP_016874649.1
XM_017019161.1 Intron XP_016874650.1
XM_017019162.1 Intron XP_016874651.1
XM_017019163.1 Intron XP_016874652.1
XM_017019164.1 Intron XP_016874653.1
XM_017019165.1 Intron XP_016874654.1
XM_017019166.1 Intron XP_016874655.1
XM_017019167.1 Intron XP_016874656.1
XM_017019168.1 Intron XP_016874657.1

View Full Product Details