Product Details

SNP ID
rs9457730
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:159799543 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTTTGAGGACCATAGTTCCAAAAC[C/G]CTGTTCAAGTAGAAGCCTAGAATTC
Phenotype
MIM: 611831 MIM: 186980
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MRPL18 PubMed Links

Gene Details

Gene
MRPL18
Gene Name
mitochondrial ribosomal protein L18
There are no transcripts associated with this gene.

Gene
PNLDC1
Gene Name
PARN like, ribonuclease domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271862.1 87 Intron NP_001258791.1
NM_173516.2 87 Intron NP_775787.1
XM_011535491.2 87 Intron XP_011533793.1
XM_011535493.2 87 Intron XP_011533795.1
XM_017010312.1 87 UTR 5 XP_016865801.1
XM_017010313.1 87 UTR 5 XP_016865802.1
XM_017010314.1 87 UTR 5 XP_016865803.1
XM_017010315.1 87 Intron XP_016865804.1
XM_017010316.1 87 Intron XP_016865805.1
Gene
TCP1
Gene Name
t-complex 1
There are no transcripts associated with this gene.

View Full Product Details