Product Details

SNP ID
rs10110868
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:28099302 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTTAATTGTTGTTGTTGTTTTGT[C/G]TAAGACAGTAGTTGTCAACCAGGGG
Phenotype
MIM: 612722
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ELP3 PubMed Links

Gene Details

Gene
ELP3
Gene Name
elongator acetyltransferase complex subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284220.1 Intron NP_001271149.1
NM_001284222.1 Intron NP_001271151.1
NM_001284224.1 Intron NP_001271153.1
NM_001284225.1 Intron NP_001271154.1
NM_001284226.1 Intron NP_001271155.1
NM_018091.5 Intron NP_060561.3
XM_006716354.2 Intron XP_006716417.1
XM_017013604.1 Intron XP_016869093.1

View Full Product Details