Product Details

SNP ID
rs9828870
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:118947162 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCCTCCAGCCCCCTAAGAACTAG[A/G]GAGGGAGCCTCACAACATTCCAATT
Phenotype
MIM: 608351
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IGSF11 PubMed Links

Gene Details

Gene
IGSF11
Gene Name
immunoglobulin superfamily member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015887.1 Intron NP_001015887.1
NM_152538.2 Intron NP_689751.2
XM_006713517.1 Intron XP_006713580.1
XM_011512462.2 Intron XP_011510764.1
XM_011512463.1 Intron XP_011510765.1
XM_011512464.2 Intron XP_011510766.1
XM_011512465.1 Intron XP_011510767.1
XM_011512466.2 Intron XP_011510768.1
XM_011512467.2 Intron XP_011510769.1
XM_011512468.1 Intron XP_011510770.1
XM_011512469.1 Intron XP_011510771.1
XM_011512470.1 Intron XP_011510772.1
XM_011512471.2 Intron XP_011510773.1
XM_011512472.1 Intron XP_011510774.1
XM_011512474.2 Intron XP_011510776.1
XM_017005789.1 Intron XP_016861278.1
XM_017005790.1 Intron XP_016861279.1
Gene
IGSF11-AS1
Gene Name
IGSF11 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details