Product Details

SNP ID
rs4952834
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:46550323 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAACCAAGGACTAAAGTCATAGCT[C/G]CTAAATAATAGCATCAGCACAACCC
Phenotype
MIM: 605857
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ATP6V1E2 PubMed Links

Gene Details

Gene
ATP6V1E2
Gene Name
ATPase H+ transporting V1 subunit E2
There are no transcripts associated with this gene.

Gene
RHOQ
Gene Name
ras homolog family member Q
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012249.3 Intron NP_036381.2
XM_005264229.2 Intron XP_005264286.1
XM_011532726.2 Intron XP_011531028.1
XM_011532728.2 Intron XP_011531030.1
XM_017003716.1 Intron XP_016859205.1

View Full Product Details