Product Details

SNP ID
rs6130935
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45792026 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCATGGGAGCCACTGGCGACGCC[A/G]AGCAGCCGCGGGGACCTAGCGGGGC
Phenotype
MIM: 611388
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNTTIP1 PubMed Links

Gene Details

Gene
DNTTIP1
Gene Name
deoxynucleotidyltransferase, terminal, interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052951.2 90 Missense Mutation AAG,GAG K8E NP_443183.1
Gene
WFDC3
Gene Name
WAP four-disulfide core domain 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080614.1 90 Intron NP_542181.1
XM_011528553.2 90 Intron XP_011526855.1
XM_017027668.1 90 Intron XP_016883157.1

View Full Product Details