Product Details

SNP ID
rs6022226
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:37988216 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCGGGTGGCACCAGGCATGGCTTC[C/T]TGAGGAGAGGCACGGCAAGTTGCTT
Phenotype
MIM: 614425
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TTI1 PubMed Links

Gene Details

Gene
TTI1
Gene Name
TELO2 interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303457.1 Intron NP_001290386.1
NM_014657.2 Intron NP_055472.1

View Full Product Details