Product Details

SNP ID
rs7513011
Assay Type
Functionally tested
NCBI dbSNP Submissions
45
Location
Chr.1:231020327 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGCCCATACCTCAGACGTTACTCA[A/T]TGAGTCCAGTGAGGATCGCATCTGT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM89A PubMed Links

Gene Details

Gene
FAM89A
Gene Name
family with sequence similarity 89 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198552.2 Intron NP_940954.1
Gene
MIR1182
Gene Name
microRNA 1182
There are no transcripts associated with this gene.

View Full Product Details