Product Details

SNP ID
rs10227997
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:92203156 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGACACAAAAAACACAACTGTGTC[C/T]CATGGCTTTTCCTATCTCTGACACT
Phenotype
MIM: 604214
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRIT1 PubMed Links

Gene Details

Gene
KRIT1
Gene Name
KRIT1, ankyrin repeat containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013406.1 Intron NP_001013424.1
NM_004912.3 Intron NP_004903.2
NM_194454.1 Intron NP_919436.1
NM_194455.1 Intron NP_919437.1
NM_194456.1 Intron NP_919438.1
XM_005250660.3 Intron XP_005250717.1
XM_005250662.3 Intron XP_005250719.1
XM_005250665.3 Intron XP_005250722.1
XM_005250666.3 Intron XP_005250723.1
XM_005250667.2 Intron XP_005250724.1
XM_005250668.3 Intron XP_005250725.1
XM_005250669.3 Intron XP_005250726.1
XM_006716161.3 Intron XP_006716224.1
XM_006716162.3 Intron XP_006716225.1
XM_006716163.3 Intron XP_006716226.1
XM_011516651.2 Intron XP_011514953.1
XM_011516653.2 Intron XP_011514955.1
XM_011516654.2 Intron XP_011514956.1
XM_011516655.2 Intron XP_011514957.1
XM_011516656.2 Intron XP_011514958.1
XM_011516657.2 Intron XP_011514959.1
XM_011516658.2 Intron XP_011514960.1
XM_011516659.2 Intron XP_011514961.1
XM_011516660.2 Intron XP_011514962.1
XM_011516661.2 Intron XP_011514963.1
XM_017012755.1 Intron XP_016868244.1
XM_017012756.1 Intron XP_016868245.1
XM_017012757.1 Intron XP_016868246.1
XM_017012758.1 Intron XP_016868247.1
XM_017012759.1 Intron XP_016868248.1
Gene
MIR1285-1
Gene Name
microRNA 1285-1
There are no transcripts associated with this gene.

View Full Product Details