Product Details

SNP ID
rs9658071
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:35347348 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAATTATTGAAAGTCTATGGCTTAC[A/G]TAGTTTTTTCAGGAGCATGGAGATT
Phenotype
MIM: 600409
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPARD PubMed Links

Gene Details

Gene
PPARD
Gene Name
peroxisome proliferator activated receptor delta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171818.1 Intron NP_001165289.1
NM_001171819.1 Intron NP_001165290.1
NM_001171820.1 Intron NP_001165291.1
NM_006238.4 Intron NP_006229.1
NM_177435.2 Intron NP_803184.1
XM_005249193.1 Intron XP_005249250.1
XM_006715120.1 Intron XP_006715183.1
XM_006715123.1 Intron XP_006715186.1
XM_011514707.1 Intron XP_011513009.1
XM_011514709.1 Intron XP_011513011.1
XM_011514710.1 Intron XP_011513012.1
XM_017010971.1 Intron XP_016866460.1
XM_017010972.1 Intron XP_016866461.1
XM_017010973.1 Intron XP_016866462.1
XM_017010974.1 Intron XP_016866463.1
XM_017010975.1 Intron XP_016866464.1

View Full Product Details