Product Details

SNP ID
rs10474578
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:79246467 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGACAGTAAGAGGTAGGATGCAAG[C/T]GAGTTTGAGACCTGACTTCTCCTTG
Phenotype
MIM: 604279
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
JMY PubMed Links

Gene Details

Gene
JMY
Gene Name
junction mediating and regulatory protein, p53 cofactor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152405.4 Intron NP_689618.4
XM_005248430.2 Intron XP_005248487.1
XM_011543155.2 Intron XP_011541457.1

View Full Product Details