Product Details

SNP ID
rs9596503
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:51338888 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCCTGAGAGATAATTATTAAGAA[C/T]TGCATTCAGTTCTTCTTTAATCTGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR5693 PubMed Links

Gene Details

Gene
MIR5693
Gene Name
microRNA 5693
There are no transcripts associated with this gene.

Gene
SERPINE3
Gene Name
serpin family E member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101320.1 3913 Intron NP_001094790.1
XM_017020707.1 3913 UTR 5 XP_016876196.1

View Full Product Details