Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001018058.2 | 3980 | UTR 3 | NP_001018068.1 | ||
NM_001244583.1 | 3980 | UTR 3 | NP_001231512.1 | ||
NM_012252.3 | 3980 | UTR 3 | NP_036384.1 | ||
XM_011515963.1 | 3980 | UTR 3 | XP_011514265.1 | ||
XM_011515964.2 | 3980 | UTR 3 | XP_011514266.1 | ||
XM_011515965.2 | 3980 | UTR 3 | XP_011514267.1 | ||
XM_011515966.2 | 3980 | UTR 3 | XP_011514268.1 | ||
XM_011515969.1 | 3980 | UTR 3 | XP_011514271.1 | ||
XM_011515970.1 | 3980 | UTR 3 | XP_011514272.1 | ||
XM_011515972.2 | 3980 | UTR 3 | XP_011514274.1 | ||
XM_017011874.1 | 3980 | UTR 3 | XP_016867363.1 | ||
XM_017011875.1 | 3980 | UTR 3 | XP_016867364.1 | ||
XM_017011876.1 | 3980 | UTR 3 | XP_016867365.1 | ||
XM_017011877.1 | 3980 | UTR 3 | XP_016867366.1 | ||
XM_017011878.1 | 3980 | UTR 3 | XP_016867367.1 |