Product Details

SNP ID
rs9261283
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30067708 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATAGGGAGGTACAGTTAGGATATA[A/G]GTGTTGCTGCTTGGGGTGGTTATGT
Phenotype
MIM: 606670 MIM: 607524 MIM: 607525 MIM: 615714
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPP1R11 PubMed Links

Gene Details

Gene
PPP1R11
Gene Name
protein phosphatase 1 regulatory inhibitor subunit 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021959.2 Intron NP_068778.1
XM_006715174.3 Intron XP_006715237.1
Gene
RNF39
Gene Name
ring finger protein 39
There are no transcripts associated with this gene.

Gene
ZNRD1
Gene Name
zinc ribbon domain containing 1
There are no transcripts associated with this gene.

Gene
ZNRD1ASP
Gene Name
zinc ribbon domain containing 1 antisense, pseudogene
There are no transcripts associated with this gene.

View Full Product Details