Product Details

SNP ID
rs10056335
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:122863483 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAATGGTGGGGTGGGAGGGCAGAG[C/T]ATAGGTCTGGTAGGCAGCAATAAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SNX24 PubMed Links

Gene Details

Gene
SNX24
Gene Name
sorting nexin 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014035.2 Intron NP_054754.1
XM_005271972.4 Intron XP_005272029.1
XM_006714592.3 Intron XP_006714655.1
XM_011543349.2 Intron XP_011541651.1
XM_011543350.2 Intron XP_011541652.1
XM_011543352.1 Intron XP_011541654.1
XM_017009394.1 Intron XP_016864883.1
XM_017009395.1 Intron XP_016864884.1
XM_017009396.1 Intron XP_016864885.1

View Full Product Details