Product Details

SNP ID
rs6025496
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:57332396 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCCTGAACATAGAGCACATTTAA[A/G]GCTTTCTCCTGACAGATCTTCTGCC
Phenotype
MIM: 605114
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPO11 PubMed Links

Gene Details

Gene
SPO11
Gene Name
SPO11, initiator of meiotic double stranded breaks
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012444.2 Intron NP_036576.1
NM_198265.1 Intron NP_937998.1

View Full Product Details