Product Details

SNP ID
rs9469419
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33394383 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGACTCTGGGTTTTTGGCTTGAGCA[A/G]CTGAAAGATGGAGTTATATCAATTG
Phenotype
MIM: 603763
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIFC1 PubMed Links

Gene Details

Gene
KIFC1
Gene Name
kinesin family member C1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002263.3 Intron NP_002254.2
XM_011514585.1 Intron XP_011512887.1
XM_011514587.2 Intron XP_011512889.1
XM_017010836.1 Intron XP_016866325.1
XM_017010837.1 Intron XP_016866326.1

View Full Product Details