Product Details

SNP ID
rs9294308
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:84671787 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTGTTTAGGTGGTGTCTTAAATTA[A/G]GCATATGGTTGGTTATGTACACACA
Phenotype
MIM: 604613
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TBX18 PubMed Links

Gene Details

Gene
TBX18
Gene Name
T-box 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080508.2 Intron NP_001073977.1
XM_006715602.3 Intron XP_006715665.2
XM_011536247.2 Intron XP_011534549.2
XM_011536248.2 Intron XP_011534550.2
XM_011536249.2 Intron XP_011534551.2
XM_017011472.1 Intron XP_016866961.1
XM_017011473.1 Intron XP_016866962.1

View Full Product Details