Product Details

SNP ID
rs4813030
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:1116966 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACGCAAAGGCCCAAACAGAACACA[C/T]GAAACAGGGAAACTGGCTGGGAAGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105372493 PubMed Links

Gene Details

Gene
LOC105372493
Gene Name
uncharacterized LOC105372493
There are no transcripts associated with this gene.

Gene
PSMF1
Gene Name
proteasome inhibitor subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323407.1 Intron NP_001310336.1
NM_001323408.1 Intron NP_001310337.1
NM_001323409.1 Intron NP_001310338.1
NM_001323410.1 Intron NP_001310339.1
NM_006814.4 Intron NP_006805.2
NM_178578.3 Intron NP_848693.2

View Full Product Details