Product Details
- SNP ID
-
hCV30024733
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:165258455 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGGGTTATTTTTAATCAATCTCAG[T/G]GTTAACACTAGTGCAACTCTGCACT
- Phenotype
-
MIM: 605774
- Polymorphism
- T/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
KLHL2
PubMed Links
Gene Details
- Gene
- KLHL2
- Gene Name
- kelch like family member 2
View Full Product Details