Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001024215.1 | Intron | NP_001019386.1 | |||
NM_001024216.1 | Intron | NP_001019387.1 | |||
NM_017556.2 | Intron | NP_060026.2 | |||
XM_005245900.1 | Intron | XP_005245957.1 | |||
XM_005245901.1 | Intron | XP_005245958.1 | |||
XM_005245902.1 | Intron | XP_005245959.1 | |||
XM_005245903.1 | Intron | XP_005245960.1 | |||
XM_005245909.3 | Intron | XP_005245966.1 | |||
XM_006710704.3 | Intron | XP_006710767.1 | |||
XM_006710705.1 | Intron | XP_006710768.1 | |||
XM_011541616.2 | Intron | XP_011539918.1 | |||
XM_011541617.2 | Intron | XP_011539919.1 | |||
XM_011541618.2 | Intron | XP_011539920.1 | |||
XM_017001519.1 | Intron | XP_016857008.1 | |||
XM_017001520.1 | Intron | XP_016857009.1 | |||
XM_017001521.1 | Intron | XP_016857010.1 | |||
XM_017001522.1 | Intron | XP_016857011.1 | |||
XM_017001523.1 | Intron | XP_016857012.1 | |||
XM_017001524.1 | Intron | XP_016857013.1 | |||
XM_017001525.1 | Intron | XP_016857014.1 | |||
XM_017001526.1 | Intron | XP_016857015.1 |