Product Details

SNP ID
rs9421246
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:116848591 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAGAACACCCTGTCCGTCGCCGCC[C/T]ACTCCTGCAGCAGAGGAAGTATCTA
Phenotype
MIM: 610701
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ENO4 PubMed Links

Gene Details

Gene
ENO4
Gene Name
enolase family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242699.1 362 Intron NP_001229628.1
XM_005269815.3 362 UTR 5 XP_005269872.1
XM_006717835.3 362 UTR 5 XP_006717898.1
XM_006717836.3 362 Intron XP_006717899.1
XM_011539797.2 362 UTR 5 XP_011538099.1
XM_011539799.2 362 UTR 5 XP_011538101.1
XM_017016241.1 362 UTR 5 XP_016871730.1
Gene
HSPA12A
Gene Name
heat shock protein family A (Hsp70) member 12A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025015.2 362 Intron NP_079291.2
XM_005269672.3 362 Intron XP_005269729.1
XM_005269673.4 362 Intron XP_005269730.1
XM_011539579.2 362 Intron XP_011537881.1
XM_011539580.2 362 Intron XP_011537882.1
XM_017016032.1 362 Intron XP_016871521.1

View Full Product Details