Product Details

SNP ID
rs6060718
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:35957920 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCACTTAGCTTCCAACACAAGGC[C/T]TGACTCCTAGGTAGCTTCAGTGTTG
Phenotype
MIM: 610335 MIM: 610416
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CNBD2 PubMed Links

Gene Details

Gene
CNBD2
Gene Name
cyclic nucleotide binding domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207076.2 Intron NP_001194005.1
NM_001304367.1 Intron NP_001291296.1
NM_080834.3 Intron NP_543024.2
XM_005260295.2 Intron XP_005260352.1
XM_011528590.2 Intron XP_011526892.1
XM_011528592.1 Intron XP_011526894.1
XM_011528593.2 Intron XP_011526895.1
XM_011528598.1 Intron XP_011526900.1
XM_017027683.1 Intron XP_016883172.1
Gene
PHF20
Gene Name
PHD finger protein 20
There are no transcripts associated with this gene.

Gene
SCAND1
Gene Name
SCAN domain containing 1
There are no transcripts associated with this gene.

View Full Product Details