Product Details

SNP ID
rs7543897
Assay Type
Functionally tested
NCBI dbSNP Submissions
60
Location
Chr.1:154706510 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGGAGCAATGAAAACATTTTTCTG[C/T]GCCGCTCATTACACTAAATCTTGAT
Phenotype
MIM: 602983
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCNN3 PubMed Links

Gene Details

Gene
KCNN3
Gene Name
potassium calcium-activated channel subfamily N member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204087.1 3976 UTR 3 NP_001191016.1
NM_002249.5 3976 UTR 3 NP_002240.3
NM_170782.2 3976 UTR 3 NP_740752.1

View Full Product Details