Product Details

SNP ID
rs2449509
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:97775805 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTCCCCGTCCCCGCCGCTGCAGC[C/G]GTCGCCTTCGGAGCGAAGGGTACCG
Phenotype
MIM: 613296
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LAPTM4B PubMed Links

Gene Details

Gene
LAPTM4B
Gene Name
lysosomal protein transmembrane 4 beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018407.4 225 Silent Mutation GCC,GCG A23A NP_060877.3

View Full Product Details