Product Details

SNP ID
rs10142846
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:35285881 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTGTGACACGATAGGTAAGAATTA[G/T]CTGCTTTCCAAATTAAGACAGAGTG
Phenotype
MIM: 602855
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PSMA6 PubMed Links

Gene Details

Gene
PSMA6
Gene Name
proteasome subunit alpha 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282232.1 Intron NP_001269161.1
NM_001282233.1 Intron NP_001269162.1
NM_001282234.1 Intron NP_001269163.1
NM_002791.2 Intron NP_002782.1
XM_017021468.1 Intron XP_016876957.1

View Full Product Details