Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_000176.2 | 6099 | UTR 3 | NP_000167.1 | ||
NM_001018074.1 | 6099 | UTR 3 | NP_001018084.1 | ||
NM_001018075.1 | 6099 | UTR 3 | NP_001018085.1 | ||
NM_001018076.1 | 6099 | UTR 3 | NP_001018086.1 | ||
NM_001018077.1 | 6099 | UTR 3 | NP_001018087.1 | ||
NM_001020825.1 | 6099 | UTR 3 | NP_001018661.1 | ||
NM_001024094.1 | 6099 | UTR 3 | NP_001019265.1 | ||
NM_001204258.1 | 6099 | UTR 3 | NP_001191187.1 | ||
NM_001204259.1 | 6099 | UTR 3 | NP_001191188.1 | ||
NM_001204260.1 | 6099 | UTR 3 | NP_001191189.1 | ||
NM_001204261.1 | 6099 | UTR 3 | NP_001191190.1 | ||
NM_001204262.1 | 6099 | UTR 3 | NP_001191191.1 | ||
NM_001204263.1 | 6099 | UTR 3 | NP_001191192.1 | ||
NM_001204264.1 | 6099 | UTR 3 | NP_001191193.1 | ||
NM_001204265.1 | 6099 | Intron | NP_001191194.1 | ||
XM_005268419.3 | 6099 | Intron | XP_005268476.1 | ||
XM_005268420.4 | 6099 | Intron | XP_005268477.1 | ||
XM_005268422.3 | 6099 | Intron | XP_005268479.1 | ||
XM_005268423.3 | 6099 | Intron | XP_005268480.1 | ||
XM_011537637.2 | 6099 | UTR 3 | XP_011535939.1 | ||
XM_017009397.1 | 6099 | UTR 3 | XP_016864886.1 | ||
XM_017009398.1 | 6099 | UTR 3 | XP_016864887.1 |