Product Details

SNP ID
rs10231092
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:76048112 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGGAGGTCGTTGGAGTCACTTCC[C/G]CGTCACCAGCTCCTGTGCCTGCCAG
Phenotype
MIM: 154100 MIM: 616695
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MDH2 PubMed Links

Gene Details

Gene
MDH2
Gene Name
malate dehydrogenase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282403.1 94 UTR 5 NP_001269332.1
NM_001282404.1 94 UTR 5 NP_001269333.1
NM_005918.3 94 UTR 5 NP_005909.2
Gene
STYXL1
Gene Name
serine/threonine/tyrosine interacting-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317785.1 94 Intron NP_001304714.1
NM_001317786.1 94 Intron NP_001304715.1
NM_001317787.1 94 Intron NP_001304716.1
NM_001317788.1 94 Intron NP_001304717.1
NM_001317789.1 94 Intron NP_001304718.1
NM_016086.2 94 Intron NP_057170.1
XM_011516292.2 94 Intron XP_011514594.1
XM_011516293.2 94 Intron XP_011514595.1
XM_011516300.1 94 Intron XP_011514602.1
XM_017012296.1 94 Intron XP_016867785.1
XM_017012297.1 94 Intron XP_016867786.1
XM_017012298.1 94 Intron XP_016867787.1
XM_017012299.1 94 Intron XP_016867788.1
XM_017012300.1 94 Intron XP_016867789.1
XM_017012301.1 94 Intron XP_016867790.1
XM_017012302.1 94 Intron XP_016867791.1

View Full Product Details