Product Details

SNP ID
rs6035841
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:21311590 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTTTTGAGTATGTACCTGTGAGTG[C/G]AATCATTGGATCATGTAATTCTGTG
Phenotype
MIM: 608851
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
XRN2 PubMed Links

Gene Details

Gene
XRN2
Gene Name
5'-3' exoribonuclease 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317960.1 Intron NP_001304889.1
NM_012255.4 Intron NP_036387.2
XM_017027722.1 Intron XP_016883211.1
XM_017027723.1 Intron XP_016883212.1
XM_017027724.1 Intron XP_016883213.1

View Full Product Details