Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001003679.3 | Intron | NP_001003679.1 | |||
NM_001003680.3 | Intron | NP_001003680.1 | |||
NM_001198687.1 | Intron | NP_001185616.1 | |||
NM_001198688.1 | Intron | NP_001185617.1 | |||
NM_001198689.1 | Intron | NP_001185618.1 | |||
NM_002303.5 | Intron | NP_002294.2 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001198681.1 | Intron | NP_001185610.1 | |||
NM_001198683.1 | Intron | NP_001185612.1 | |||
NM_017526.4 | Intron | NP_059996.1 |