Product Details

SNP ID
rs10250243
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:132796195 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAGTTCAGGGCAAGTGGAAAAAA[G/T]TATAATTGTAGTAGAATCCTTTTTT
Phenotype
MIM: 613748
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CHCHD3 PubMed Links

Gene Details

Gene
CHCHD3
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317177.1 Intron NP_001304106.1
NM_001317178.1 Intron NP_001304107.1
NM_017812.3 Intron NP_060282.1

View Full Product Details