Product Details

SNP ID
hCV30069264
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:1351581 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTAGCATCTGAGCTGGGGATGACC[A/G]TTACGAGTCCACAGCTGTTTGACCA
Phenotype
MIM: 605066
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
YWHAE PubMed Links

Gene Details

Gene
YWHAE
Gene Name
tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein epsilon
There are no transcripts associated with this gene.

View Full Product Details