Product Details

SNP ID
rs6854081
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:122895552 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATAAGGCTCAGCTGCTTTATAGTTC[G/T]CTGGCTAATGCAGCAGATACTCTTC
Phenotype
MIM: 134920 MIM: 606261
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FGF2 PubMed Links

Gene Details

Gene
FGF2
Gene Name
fibroblast growth factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002006.4 4091 UTR 3 NP_001997.5
Gene
NUDT6
Gene Name
nudix hydrolase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007083.4 4091 Intron NP_009014.2
NM_198041.2 4091 Intron NP_932158.1

View Full Product Details