Product Details
- SNP ID
-
rs9476507
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:14119976 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGGATGTCAGCTGGTGACGTAACT[C/T]GGTAACGCATGAGCTTGTTTATTGG
- Phenotype
-
MIM: 604534
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CD83
PubMed Links
Gene Details
- Gene
- CD83
- Gene Name
- CD83 molecule
View Full Product Details