Product Details

SNP ID
rs10409169
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:20083272 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTAGTCCCACCCGATGAGAAATAC[C/T]CACAGGTGTGGAGGGGCAGGCCACC
Phenotype
MIM: 603973
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF90 PubMed Links

Gene Details

Gene
ZNF90
Gene Name
zinc finger protein 90
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007138.1 Intron NP_009069.1

View Full Product Details