Product Details
- SNP ID
-
rs4519019
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:1070354 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCTTCCAGTTTTCTAACATTTTTGG[A/G]TTATGCTTGGCTTTTAAAAAAGTTT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
WDR37
PubMed Links
Gene Details
- Gene
- WDR37
- Gene Name
- WD repeat domain 37
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014023.3 |
|
Intron |
|
|
NP_054742.2 |
View Full Product Details