Product Details

SNP ID
rs10521180
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:11247576 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGGAACCACAAAATGATGGTTTCA[A/G]TTTACAAGCATGTAAGATGTCTCAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SHISA6 PubMed Links

Gene Details

Gene
SHISA6
Gene Name
shisa family member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001173461.1 Intron NP_001166932.1
NM_001173462.1 Intron NP_001166933.1
NM_207386.3 Intron NP_997269.2
XM_017024618.1 Intron XP_016880107.1

View Full Product Details